Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Bovine IgM (H+L) Secondary Antibody (IL-A30), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB555790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB555790 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB555790 Supplier Novus Biologicals Supplier No. NBP268419FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgM (H+L) Monoclonal specifically detects IgM (H+L) in Bovine samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA).

Specifications

Antigen IgM (H+L)
Applications ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone IL-A30
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Bovine peripheral blood mononuclear cells depleted of T lymphocytes
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.