Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Canine IgA/IgG/IgM (H+L) Secondary Antibody (K99 2E3), Biotin, Novus Biologicals™
SDP

Catalog No. NB523912 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB523912 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB523912 Supplier Novus Biologicals Supplier No. NBP311626B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgA/IgG/IgM (H+L) Monoclonal antibody specifically detects IgA/IgG/IgM (H+L) in Canine samples. It is validated for ELISA

Specifications

Antigen IgA/IgG/IgM (H+L)
Applications ELISA
Classification Monoclonal
Clone K99 2E3
Conjugate Biotin
Formulation PBS
Host Species Mouse
Immunogen Canine immunoglobulin
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.