Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Canine IgA/IgG/IgM (H+L) Secondary Antibody (K99 2E3), Novus Biologicals™

Mouse Monoclonal Antibody
$149.80 - $333.00
Specifications
Antigen | IgA/IgG/IgM (H+L) |
---|---|
Clone | K99 2E3 |
Dilution | ELISA 1:100 - 1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IgA/IgG/IgM (H+L) Monoclonal antibody specifically detects IgA/IgG/IgM (H+L) in Canine samples. It is validated for ELISASpecifications
IgA/IgG/IgM (H+L) | |
ELISA 1:100 - 1:10000 | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
IgG1 | |
Protein A purified |
K99 2E3 | |
ELISA | |
Unconjugated | |
Mouse | |
Canine | |
Canine immunoglobulin | |
Secondary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title