Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Canine IgA/IgG/IgM (H+L) Secondary Antibody (K99 2E3), R&D Systems™
SDP

Catalog No. NB397971 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397971 0.025 mg
NB397970 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397971 Supplier R&D Systems Supplier No. NBP3116260.025MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgA/IgG/IgM (H+L) Monoclonal specifically detects IgA/IgG/IgM (H+L) in Canine samples. It is validated for ELISA.

Specifications

Antigen IgA/IgG/IgM (H+L)
Applications ELISA
Classification Monoclonal
Clone K99 2E3
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:10000
Formulation PBS
Host Species Mouse
Immunogen Canine immunoglobulin
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Canine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.