Learn More
Mouse anti-Canine IgG2 Heavy Chain Secondary Antibody (CA4F1), DyLight 550, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG2 Heavy Chain |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | CA4F1 |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Canine IgG |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.