Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine parvovirus, Mouse, Clone: 5G7, Abnova™

Catalog No. 89134719 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-719 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-719 Supplier Abnova Corporation Supplier No. MAB0589
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against canine parvovirus.

Specifications

Antigen Canine parvovirus
Applications ELISA, Inhibition Assays
Classification Monoclonal
Clone 5G7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against canine parvovirus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Canine parvovirus.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.