Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carcino Embryonic Antigen Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89134953 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-953 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-953 Supplier Abnova Corporation Supplier No. MAB1395
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Carcino Embryonic Antigen.

Specifications

Antigen Carcino Embryonic Antigen
Applications Immunohistochemistry
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Carcino Embryonic Antigen.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified human Carcino Embryonic Antigen.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to human CEA. No cross-reactivity with human NCA, NCA2 or BGP1 (Biliary glycoprotein-l).
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.