Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR-alpha/beta Mouse anti-Human, Unlabeled, Clone: WT31, BD

Catalog No. BDB347770 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347770 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347770 Supplier BD Biosciences Supplier No. 347770
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone WT31 is derived from hybridization of mouse Sp2/0-Ag14 myeloma cells with spleen cells from BALB/c mice immunized with human thymocytes.

WT31 (Anti–TCR-α/β-1) recognizes a conformational epitope formed by the T-cell receptor (TCR) for antigen and the CD3 epsilon chain. The α/β TCR is a disulfide-linked 80-kdalton (kDa) heterodimer consisting of a 44-kDa α chain and a 37-kDa β chain.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR-alpha/beta
Applications Flow Cytometry
Classification Monoclonal
Clone WT31
Concentration 6.25μg/mL
Conjugate Unconjugated
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human thymocytes
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6955 6957
Target Species Human
Content And Storage Store at 2-8°C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.