Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD70 Mouse anti-Human, BB515, Clone: Ki-24, BD

Catalog No. BDB565156 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565156 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565156 Supplier BD Biosciences Supplier No. 565156
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Ki-24 monoclonal antibody specifically binds to human CD70. CD70 is a type II transmembrane glycoprotein and member of the TNF Superfamily. CD70 is also known as Tumor necrosis factor ligand superfamily member 7 (TNFSF7), CD27 ligand (CD27-L, CD27L, CD27LG), and KI-24 antigen. The CD70 antigen immunoprecipitates as five bands (50, 70, 90, 100 and 160 kDa) under non-reducing conditions. CD70 is strongly expressed on Reed-Sternberg cells, some activated T or B cells and Epstein Barr Virus (EBV)-positive lymphoblastoid cell lines. CD70 plays roles in the activation, proliferation and differentiation of B cells and T cells including the enhanced production of cytotoxic T cells.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD70
Applications Flow Cytometry
Classification Monoclonal
Clone Ki-24
Conjugate Brilliant Blue 515 (BB515)
Description CDw70; CD27 ligand; CD27-L; CD27L; CD27LG; Ki-24 antigen; TNFSF7
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG3 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.