Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cdc6, Mouse, Clone: Cdc6 9H8/5, Abnova™

Catalog No. 89113155 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-155 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-155 Supplier Abnova Corporation Supplier No. MAB7254
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length cdc6.

Specifications

Antigen cdc6
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Cdc6 9H8/5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length cdc6.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene cdc6
Gene Alias mis10/pol3/pold
Gene Symbols cdc6
Host Species Mouse
Immunogen Full length yeast cdc6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2540255
Test Specificity Cdc6 (Cdc6 9H8/5). In WB predicted molecular weight 58 KDa.
Target Species Yeast
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.