Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH22, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107282 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-282 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-282 Supplier Abnova Corporation Supplier No. H00064405A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CDH22.

This gene is a member of the cadherin superfamily. The gene product is composed of five cadherin repeat domains and a cytoplasmic tail similar to the highly conserved cytoplasmic region of classical cadherins. Expressed predominantly in the brain, this putative calcium-dependent cell adhesion protein may play an important role in morphogenesis and tissue formation in neural and non-neural cells during development and maintenance of the brain and neuroendocrine organs. [provided by RefSeq

Sequence: PPRFPQKMYQFSIQESAPIGTAVGRVKAEDSDVGENTDMTYHLKDESSSGGDVFKVTTDSDTQEAIIVVQKRLDFESQPVHTVILEALNKFVDPRFADLGTFRDQAIVRV

Specifications

Antigen CDH22
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CDH22.
Formulation 50% glycerol
Gene CDH22
Gene Accession No. NM_021248
Gene Alias C20orf25/MGC39564/dJ998H6.1
Gene Symbols CDH22
Host Species Mouse
Immunogen CDH22 (NP_067071, 274 a.a. to 383 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64405
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.