Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP7, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-493 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-493 Supplier Abnova Corporation Supplier No. H00091782B01P
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CHMP7 protein.

Sequence: MWSPEREAEAPAGGDPAGLLPPEWEEDEERMSFLFSAFKRSREVNSTDWDSKMGFWAPLVLSHSRRQGVVRLRLRDLQEAFQRKGSVPLGLATVLQDLLRRGELQRESDFMASVDSSWISWGVGVFLLKPLKWTLSNMLGDNKVPAEEVLVAVELLKEKAEEVYRLYQNSPLSSHPVVALSELSTLCANSCPDERTFYLVLLQLQKEKRVTVLEQNGEKIVKFARGPRAKVSPVNDVDVGVYQLMQSEQLLSRKVESLSQEAERCKEEARRACRAGKKQLALRSLKAKQRTEKRIEALHAKLDTVQGILDRIYASQTDQMVFNAYQAGVGALKLSMKDVTVEKAESLVDQIQELCDTQDEVSQTLAGGVTNGLDFDSEELEKELDILLQDTTKEPLDLPDNPRNRHFTNSVPNPRISDAELEAELEKLSLSEGGLVPSSKSPKRQLEPTLKPL

Specifications

Antigen CHMP7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CHMP7 protein.
Formulation PBS with no preservative; pH 7.4
Gene CHMP7
Gene Accession No. NM_152272.2
Gene Alias MGC29816
Gene Symbols CHMP7
Host Species Mouse
Immunogen CHMP7 (NP_689485.1,1 a.a. ∽ 453 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91782
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.