Learn More
Immunology Consultants Monoclonal Mouse anti-CHO Phospholipase B-like 2 protein (PLBL2) Antibody, 0.5 mg
SDP
Supplier:  Immunology Consultants MPLB65ALY4D5
Encompass_Preferred
This mouse IgG1 kappa is purified using salt fractionation procedures and Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with CHO Phospholipase B-Like 2 (PLBL2) of recombinant origin and PLBL2 secreted by a null CHO cell line as determined by ELISA and western blot. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8914
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.