Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL11A2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104307 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-307 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-307 Supplier Abnova Corporation Supplier No. H00001302A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant COL11A2.

This gene encodes one of the two alpha chains of type XI collagen, a minor fibrillar collagen. It is located on chromosome 6 very close to but separate from the gene for retinoid X receptor beta. Type XI collagen is a heterotrimer but the third alpha chain is a post-translationally modified alpha 1 type II chain. Proteolytic processing of this type XI chain produces PARP, a proline/arginine-rich protein that is an amino terminal domain. Mutations in this gene are associated with type III Stickler syndrome, otospondylomegaepiphyseal dysplasia (OSMED syndrome), Weissenbacher-Zweymuller syndrome, autosomal dominant nonsyndromic sensorineural type 13 deafness (DFNA13), and autosomal recessive nonsyndromic sensorineural type 53 deafness (DFNB53). Alternative splicing results in multiple transcript variants. A related pseudogene is located nearby on chromosome 6. [provided by RefSeq

Sequence: APPVDVLRALRFPSLPDGVRRAKGICPADVAYRVARPAQLSAPTRQLFPGGFPKDFSLLTVVRTRPGLQAPLLTLYSAQGVRQLGLELGRPVRFLYEDQT

Specifications

Antigen COL11A2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant COL11A2.
Formulation 50% glycerol
Gene COL11A2
Gene Accession No. NM_080680
Gene Alias DFNA13/DFNB53/HKE5/PARP/STL3
Gene Symbols COL11A2
Host Species Mouse
Immunogen COL11A2 (NP_542411, 29 a.a. to 128 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1302
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.