Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRABP2, Mouse, Clone: AT2E11, Abnova™

Catalog No. 89108321 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-321 Supplier Abnova Corporation Supplier No. MAB10007
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant CRABP2.

A number of specific carrier proteins for members of the vitamin A family have been discovered. Cellular retinoic acid binding proteins (CRABP) are low molecular weight proteins whose precise function remains unknown. The inducibility of the CRABP2 gene suggests that this isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. It has been postulated that the CRABP2 gene is transcriptionally regulated by a newly synthesized regulatory protein. [provided by RefSeq

Specifications

Antigen CRABP2
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT2E11
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant CRABP2.
Dilution Western Blot (1:250-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CRABP2
Gene Alias CRABP-II/RBP6
Gene Symbols CRABP2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CRABP2.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1382
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.