Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBH, Mouse, Clone: DBH, Abnova™

Catalog No. 89136035 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-035 Supplier Abnova Corporation Supplier No. MAB5671
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against DBH.

Specifications

Antigen DBH
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone DBH41
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against DBH.
Dilution Immunohistochemistry (1:1000) Immunocytochemistry (1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Host Species Mouse
Immunogen Dopamine beta-hydroxylase (DBH).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody specifically recognizes two bands of molecular weight 70 and 75 KDa, corresponding to the soluble and membrane bound forms of the enzyme, respectively. DBH antibody specifically stains DBH containing neurons and fibers in the rat central nervous system (CNS).
Target Species Rat
Content And Storage Store at 4°C or -20°C on dry atmosphere. After reconstitution with water and glycerol (1:1), store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.