Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fcamr, Mouse, Clone: TX61, Abnova™

Catalog No. 89112948 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-948 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-948 Supplier Abnova Corporation Supplier No. MAB6585
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Fcamr.

Specifications

Antigen Fcamr
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone TX61
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Fcamr.
Dilution Immunoprecipitation (5-10 ug/mL) Flow Cyt (0.5-1 ug/mL) Immunoflourescence (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2% Block Ace)
Gene Fcamr
Gene Alias MGC129330/MGC129331
Gene Symbols Fcamr
Host Species Mouse
Immunogen Native from Mouse Fcamr expressing cell line.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64435
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.