Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mouse anti-Feline Ig Mix Secondary Antibody
Mouse Monoclonal Secondary Antibody
Supplier: Invitrogen™ SA510354
This item is not returnable.
View return policy
Description
SA1-10643 detects feline Immunoglobulin. Does not react with bovine, equine, canine, rabbit, sheep, porcine, donkey, guinea pig, goat or human. SA1-10643 has been successfully used in ELISA applications. The SA1-10643 immunogen is purified feline immunoglobulin.
Anti-Feline secondary antibodies are affinity-purified antibodies with well-characterized specificity for feline immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Feline Ig Mix | |
Monoclonal | |
0.6 mg/mL | |
PBS with no preservative; pH 7.4 | |
Purified feline immunoglobulin. | |
100 μg | |
Secondary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
FIG11-207.2 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Feline | |
Secondary Antibody | |
IgG1 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'
missing translation for 'spotOppurtunityImprovement'missing translation for 'shareContentCorrection'