Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCG, Mouse, Clone: HGL-B5, Abnova™

Catalog No. 89113474 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-474 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-474 Supplier Abnova Corporation Supplier No. MAB8497
This item is not returnable. View return policy

Mouse monoclonal antibody raised against full-length GCG.

The protein encoded by this gene is actually a preproprotein that is cleaved into four distinct mature peptides. One of these, glucagon, is a pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis. Glucagon is a ligand for a specific G-protein linked receptor whose signalling pathway controls cell proliferation. Two of the other peptides are secreted from gut endocrine cells and promote nutrient absorption through distinct mechanisms. Finally, the fourth peptide is similar to glicentin, an active enteroglucagon. [provided by RefSeq

Specifications

Antigen GCG
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone HGL-B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length GCG.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene GCG
Gene Alias GLP1/GLP2/GRPP
Gene Symbols GCG
Host Species Mouse
Immunogen Full length (31 amino acids) synthetic human GCG.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2641
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.