Learn More
Mouse Anti-Hamster Ig, κ Light Chain, Clone: HIG-29, Secondary Antibody, BD
Supplier: BD Biosciences 550636
Description
Based on ELISA, the HIG-29 antibody reacts specifically with both Syrian and Armenian hamster Ig, κ light chains. It does not react with Ig, κ light chains from other species, nor with hamster Ig heavy chains.
Host Species: Mouse
Clone: HIG-29
Isotype: IgG1 κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian Hamster IgG mAb
ELISA, Flow Cytometry

Specifications
Mouse Anti-Hamster Ig, κ Light Chain | |
HIG-29 | |
Biotin | |
0.5 mg | |
Immunology | |
Hamster |
Monoclonal | |
0.5mg/mL | |
Mouse | |
RUO | |
Secondary | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.