Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Hamster IgM, Clone: G188-2, Secondary Antibody, BD
Supplier: BD Biosciences 554031
Description
Based on ELISA, the G188-2 antibody reacts specifically with hamster IgM. The G188-2 mAb does not react with hamster IgG
Host Species: Mouse
Clone: G188-2
Isotype: IgG1, κ
Species Reactivity: Hamster
Immunogen: Armenian Hamster IgM (mAb HM40-3)
Flow Cytometry

Specifications
Mouse anti-Hamster IgM | |
G188-2 | |
Mouse | |
0.5 mg | |
Immunology | |
Hamster |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction