Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DR/HLA-DP, Mouse, Clone: MEM-136, Abnova™

Catalog No. 89108245 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-245 Supplier Abnova Corporation Supplier No. MAB0917
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native HLA-DR/HLA-DP.

Specifications

Antigen HLA-DR/HLA-DP
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MEM-136
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native HLA-DR/HLA-DP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Alias DPB1/HLA-DP1B
Gene Symbols HLA-DPB1
Host Species Mouse
Immunogen Native purified HLA-DR/HLA-DP from PHA-activated peripheral blood leucocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3115/3123/3125/3126/3127
Test Specificity This antibody recognizes common epitope on beta-chain of human HLA-DR and HLA-DP. It reacts with alpha/beta dimer as well as with dissociated beta-subunit.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.