Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-HOIL-1, Clone: 2E2,

Catalog No. MABC576MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC576MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC576MI Supplier MilliporeSigma™ Supplier No. MABC576
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects HOIL-1 clone: 2E2 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

RanBP-type and C3HC4-type zinc finger-containing protein 1 (UniProt Q9BYM8; also known as HOIL-1, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, RBCC protein interacting with PKC1, RING finger protein 54, Ubiquitin-conjugating enzyme 7-interacting protein 3) is encoded by the RBCK1 gene (also known as C20orf18, HOIL1, PBMEI, RBCK2, RNF54, UBCE7IP3, XAP3, XAP4, ZRANB4) (Gene ID 10616) in human. The linear ubiquitin assembly complex (LUBAC) is comprised of two RING-IBR-RING (RBR)-containing E3 ligases, HOIL-1L and HOIP. LUBAC conjugates a head-to-tail-linked linear polyubiquitin chain onto lysine residues on substrate proteins. Ovarian tumor deubiquitinating enzyme (DUB) with linear linkage specificity (OTULIN/Gumby/FAM105B) and cylindromatosis (CYLD) are two DUBs that catalyze the cleavage linear polyubiquitin chains. OTULIN and CYLD interact with the N-terminal PNGase/UBA or UBX (PUB) domain of HOIP and negatively regulate LUBAC-mediated cellular processes. LUBAC is reported to suppress IFN-dependent antiviral activity by mediating TRIM25 degradation and competing against TRIM25 for RIG-I interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOIL-1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2E2
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9BYM8
Gene Symbols RBCK1; C20orf18; RNF54; UBCE7IP3; XAP3; XAP4
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human HOIL-1 near the C-terminus.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_006453
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.