Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 Mouse anti-Human, Brilliant Violet 421, Clone: LB-2, BD Optibuild™

Catalog No. BDB740094 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740094 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740094 Supplier BD Biosciences Supplier No. 740094
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The LB-2 monoclonal antibody specifically binds to CD54 which is also known as Intercellular adhesion molecule-1 (ICAM-1). CD54 is an 85-110 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin supergene family. A soluble form of CD54 can also be found in biological fluids. CD54 is expressed on endothelial cells and both resting (weak) and activated (moderate) T and B lymphocytes and monocytes. CD54 is expressed on human myeloma cells, Burkitt's lymphoma cells, erythroleukemia cells, and B-lymphoblastoid cells. CD54 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18) and can act as a receptor for Rhinoviruses. The CD54 adhesion molecule plays roles in inflammatory and immune responses and neoplasia.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54
Applications Flow Cytometry
Classification Monoclonal
Clone LB-2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Human ICAM-1, HICAM-1; BB2; P3.58
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Major histocompatibility Class II antigen negative variant of human B-lymphoblastoid cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.