Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgE Heavy Chain Secondary Antibody (E454 (5D5)), PE, R&D Systems™
SDP

Catalog No. NB552549 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB552549 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB552549 Supplier R&D Systems Supplier No. NBP268297PE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgE Heavy Chain Monoclonal specifically detects IgE Heavy Chain in Human samples. It is validated for ELISA.

Specifications

Antigen IgE Heavy Chain
Applications ELISA
Classification Monoclonal
Clone E454 (5D5)
Conjugate PE
Formulation PBS
Gene Alias Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon
Host Species Mouse
Immunogen Immunoglobulin E from human serum
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.