Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgE Heavy Chain Secondary Antibody (E454 (5D5)), R&D Systems™
SDP

Catalog No. NB002821 Shop All Novus Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002821 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002821 Supplier R&D Systems Supplier No. NBP268297200UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgE Monoclonal specifically detects IgE in Human samples. It is validated for ELISA.

Specifications

Antigen IgE
Applications ELISA
Classification Monoclonal
Clone E454 (5D5)
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS
Gene Alias IgE, immunoglobulin heavy constant epsilon
Gene Symbols IGHE
Host Species Mouse
Immunogen Immunoglobulin E from human serum
Quantity 200 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Gene ID (Entrez) 3497
Test Specificity Mouse anti Human IgE antibody, clone E454 (5D5), recognizes human immunoglobulin E heavy chains, binding to an epitope expressed on the Ce2 domain.
Target Species Human
Content And Storage Store at 4°C.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.