Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG (CH3 domain) Secondary Antibody, Invitrogen™

Catalog No. MA190268
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA190268 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA190268 Supplier Thermo Scientific Supplier No. MA190268

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Secondary Antibody

MA1-90268 detects IgG CH3 domain from human samples.

Human IgG (CH3 domain) Monoclonal antibody specifically detects Human IgG (CH3 domain) in Human samples. It is validated for ELISA, Hemagglutination Assay, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG (CH3 domain)
Applications ELISA, Hemagglutination Assay, Western Blot
Classification Monoclonal
Clone A57H
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen pFc' from purified protein of Heavy Chain Disease
Purification Method purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4° C, do not freeze
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.