Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-Human IgG, Clone: DD7, Secondary Antibody,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ CBL600
Description
Specifically detects IgG Clone: DD7 in Human samples, and it is validated for Flow Cytometry

Specifications
IgG | |
Monoclonal | |
Unconjugated | |
Mouse | |
RUO | |
Secondary | |
Human | |
Purified |
Flow Cytometry | |
DD7 | |
Each isotype control is presented in phosphate buffered saline, containing 10mM sodium azide and 1mg/mL bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application. | |
1 mL | |
Secondary, Control Antibodies, Epitope Tags, General Use | |
This isotype control shows negligible cross-reactivity with human cell surface antigens on tissue sections or in cellular preparations. It therefore enables the estimation of non-specific binding of mouse monoclonals to cell surface antigens. | |
For use within 1 month of purchase store at 4°C, for long term storage aliquot antibody into small volumes and store at -20°C for up to one year | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction