Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG F(ab) Secondary Antibody (4A11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$181.18 - $332.50

Specifications

Antigen IgG F(ab)
Clone 4A11
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA 1:2000
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB500467SS
SDP
View Documents
Novus Biologicals
NB500467SS
0.02 mg
Each for $181.18
Only null left
Add to Cart
 
NB500467
SDP
View Documents
Novus Biologicals
NB500467
0.1 mg
Each for $332.50
Only null left
Add to Cart
 
Description

Description

IgG F(ab) Monoclonal specifically detects IgG F(ab) in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

IgG F(ab)
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA
Unconjugated
Mouse
Human
3500
Purified human IgG.
Secondary
Store at 4C. Do not freeze.
4A11
Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA 1:2000
Monoclonal
Purified
RUO
immunoglobulin heavy constant gamma 1 (G1m marker)
IGHG1
IgG2b
Protein A purified
The antibody 4A11 reacts with Fab fragment of human IgG.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.