Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG Fc Secondary Antibody (3D3cc), R&D Systems™
SDP

Catalog No. NB1108347 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1108347 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1108347 Supplier R&D Systems Supplier No. NB1108347
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG Fc Monoclonal specifically detects IgG Fc in Human samples. It is validated for Immunodiffusion, Sandwich ELISA, Western Blot.

Specifications

Antigen IgG Fc
Applications Immunodiffusion, Sandwich ELISA, Western Blot
Classification Monoclonal
Clone 3D3
Conjugate Unconjugated
Dilution Sandwich ELISA
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity
Primary or Secondary Secondary
Test Specificity Class-specific for human IgG. No allotype restriction. Fc-region specific. Relatively high affinity. No cross-reactivity with human IgA, IgM, IgE.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.