Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG Fc Secondary Antibody (EM-07), HRP, Novus Biologicals™
SDP

Catalog No. NBP178623H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178623H 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178623H Supplier Novus Biologicals Supplier No. NBP178623H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG Fc Monoclonal specifically detects IgG Fc in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen IgG Fc
Applications ELISA, Western Blot
Classification Monoclonal
Clone EM-07
Concentration 1 mg/ml
Conjugate HRP
Dilution Western Blot 1:1000, ELISA 1:2000
Host Species Mouse
Immunogen Fusion protein of human IgG Fc fragment.
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity This secondary antibody reacts with Fc part of human IgG heavy chain and with isolated Fc fragments.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.