Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG Fc Secondary Antibody, HRP, Invitrogen™

Catalog No. 01672143
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-143 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-143 Supplier Thermo Scientific Supplier No. SA135470

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Secondary Antibody

This antibody does not cross react with albumin fractions of human serum, human IgA, human IgM or albumin fractions from other species

Human IgG Fc Monoclonal antibody specifically detects Human IgG Fc in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG Fc
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone EFE 565
Concentration 1.2 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol, 5mg/mL BSA and no preservative; pH 7.5
Host Species Mouse
Immunogen Purified human IgG.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.