Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG Fc Secondary Antibody (SB2H2), Biotin, Novus Biologicals™
SDP

Catalog No. NB554830 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554830 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554830 Supplier Novus Biologicals Supplier No. NBP268422B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG Fc Monoclonal specifically detects IgG Fc in Human samples. It is validated for ELISA.

Specifications

Antigen IgG Fc
Applications ELISA
Classification Monoclonal
Clone SB2H2
Conjugate Biotin
Formulation PBS
Host Species Mouse
Immunogen Human IgG
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.