Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG Fc Secondary Antibody (SB2H2) [Janelia Fluor 525], R&D Systems™
SDP

Catalog No. NB788964
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB788964 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB788964 Supplier R&D Systems Supplier No. NBP268422JF525

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG Fc Monoclonal antibody specifically detects IgG Fc in Human samples. It is validated for ELISA

Specifications

Antigen IgG Fc
Applications ELISA
Classification Monoclonal
Clone SB2H2
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Human IgG
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.