Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG Fc Secondary Antibody (SB2H2), Novus Biologicals™
SDP

Catalog No. NB002833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002833 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002833 Supplier Novus Biologicals Supplier No. NBP268422200UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG Monoclonal specifically detects IgG in Human samples. It is validated for ELISA.

Specifications

Antigen IgG
Applications ELISA
Classification Monoclonal
Clone SB2H2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation TBS
Gene Symbols IGHG1
Host Species Mouse
Immunogen Human IgG
Quantity 200 μg
Gene ID (Entrez) 3500
Test Specificity Mouse anti Human IgG antibody, clone SB2H2, recognizes the Fc region of human IgG chains. The antibody cross-reacts strongly with rabbit IgG.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.