Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), Allophycocyanin/Cy7, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4D2D9G8 |
| Conjugate | APC-Cyanine7 |
| Dilution | Western Blot : 1:500 - 1:2000, ELISA : 1:10000 |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Human IgG was isolated from human sera and purified by chromatography. |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.