Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), Biotin, R&D Systems™
SDP

Catalog No. NB018684 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018684 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB018684 Supplier R&D Systems Supplier No. NBP151523B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG (H+L) Monoclonal specifically detects IgG (H+L) in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4D2D9G8
Conjugate Biotin
Dilution Western Blot : 1:500 - 1:2000, ELISA : 1:10000
Formulation PBS
Host Species Mouse
Immunogen Human IgG was isolated from human sera and purified by chromatography.
Purification Method Protein G purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.