Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG (H+L) Secondary Antibody, FITC, Invitrogen™

Catalog No. 01672142
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-142 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-142 Supplier Thermo Scientific Supplier No. SA135469

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Mouse Monoclonal Secondary Antibody

This antibody does not cross-react with HSA or other albumins.

Human IgG (H+L) Monoclonal antibody specifically detects Human IgG (H+L) in Human samples. It is validated for ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG (H+L)
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone EFE-565
Concentration 1.2 mg/mL
Conjugate FITC
Formulation PBS with 50% glycerol, 5mg/mL BSA and no preservative; pH 7.5
Host Species Mouse
Immunogen Purified human IgG.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20° C, store in dark
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.