Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Human IgG Secondary Antibody (MR12) (BSA Free), Novus Biologicals™

Mouse Monoclonal Antibody
$184.50
Specifications
Antigen | IgG |
---|---|
Clone | MR12 |
Concentration | 1 mg/mL |
Dilution | Immunohistochemistry 1:10 - 1:500 |
Applications | Immunohistochemistry |
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for ImmunohistochemistrySpecifications
IgG | |
1 mg/mL | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Immunoglobulin G, ImmunoglobulinG | |
Immunoglobulin purified from rabbit serum | |
Secondary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
MR12 | |
Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
3500 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title