Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG1 Fc Secondary Antibody, Biotin, Invitrogen™

Catalog No. 501137485
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-7485 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-7485 Supplier Invitrogen Supplier No. MH1515

Mouse Monoclonal Secondary Antibody

Human IgG1 Fc Monoclonal antibody specifically detects Human IgG1 Fc in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG1 Fc
Applications Flow Cytometry
Classification Monoclonal
Clone HP6070
Concentration 0.3-1 mg/mL
Conjugate Biotin
Formulation PBS with BSA and 5mM sodium azide; pH 7.5
Host Species Mouse
Immunogen IgG gamma 1
Purification Method purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.