Learn More
Description
Specifications
Specifications
| Antigen | IgG2 Fc |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 52G1 |
| Conjugate | Unconjugated |
| Dilution | ELISA assay dependent |
| Formulation | PBS (pH 7.4) with 0.09% Sodium Azide |
| Gene Symbols | IGHG2 |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human IgG2 from myeloma patient serum. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
