Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG3 Heavy Chain Secondary Antibody (5G12cc) [Janelia Fluor 525], Novus Biologicals™
SDP

Catalog No. NB789800 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB789800 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB789800 Supplier Novus Biologicals Supplier No. NB1001599JF525
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG3 Heavy Chain Monoclonal antibody specifically detects IgG3 Heavy Chain in Human samples. It is validated for ELISA

Specifications

Antigen IgG3 Heavy Chain
Applications ELISA
Classification Monoclonal
Clone 5G12cc
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG3 from human serum.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.