Learn More
Mouse anti-Human IgG3 (Hinge region) Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Supplier: Thermo Scientific MA134436
Description
MA1-34436 detects IgG3 hinge region from human samples. MA1-34436 has been successfully used in ELISA, flow cytometry, immunohistochemistry (frozen tissue) applications. The MA1-34436 immunogen is: Purified human IgG3 myeloma protein Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Human IgG3 (Hinge region) | |
Monoclonal | |
1 mg/mL | |
Mouse | |
Protein A | |
RUO | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen) | |
HP6050 | |
Unconjugated | |
Purified human IgG3 myeloma protein | |
100 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.