Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG4 Mouse anti-Human, Clone: HP6025, Secondary Antibody, MilliporeSigma™

Catalog No. MAB1313MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1313MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1313MI Supplier MilliporeSigma Supplier No. MAB1313

Mouse Monoclonal Antibody

Mouse anti-Human IgG4 Antibody, clone HP6025, Fc detects level of Human IgG4 and has been published and validated for use in ELISA, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Human IgG4 Fc
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone HP6025
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.2Mphosphate buffer, 0.25MNaCl, pH 7.6, with 0.1% sodium azide.
Gene Accession No. NM_001002273.1; NM_001002274.1; NM_001002275.1; NM_004001.3
Host Species Mouse
Quantity 500 μg
Primary or Secondary Secondary
Gene ID (Entrez) FCGR2B; CDw32; CD32B; IGFR2; CD32; Fc-gamma-RIIb; FCG2; FcRII-b; FCGR2
Target Species Human
Content And Storage 4°C. Do not Freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.