Learn More
Description
Specifications
Specifications
| Antigen | IgM Fc |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | CH2 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Purified human IgM. |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
