Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgM Fc Secondary Antibody (CH2), FITC, R&D Systems™
SDP

Catalog No. NBP144935 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144935 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144935 Supplier R&D Systems Supplier No. NBP144935
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgM Fc Monoclonal specifically detects IgM Fc in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IgM Fc
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone CH2
Concentration 1 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1-5 ug/ml
Gene Symbols IGHM
Host Species Mouse
Immunogen Purified human IgM.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Test Specificity This antibody reacts with Fc fragment of human IgM.
Target Species Human
Content And Storage Store at 4C in the dark.
Product Type Antibody
Isotype IgG1
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.