Learn More
Mouse anti-Human IgM Secondary Antibody, HRP, Invitrogen™
Description
This antibody does not cross react with albumin fractions of human serum, human IgA, human IgG or albumin fractions from other species
Specifications
Specifications
| Antigen | Human IgM |
| Applications | Dot Blot, ELISA, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | ICL-931 |
| Concentration | 1.2 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 50% glycerol, 5mg/mL BSA and no preservative; pH 7.5 |
| Host Species | Mouse |
| Immunogen | Purified human IgM. |
| Purification Method | Ion-exchange chromatography, Size-exclusion - Gel filtration |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.