Learn More
IL-10Rbeta (CD210b) Mouse anti-Human, Brilliant Violet 421, Clone: 90220, BD Optibuild™
Shop All BD Biosciences ProductsDescription
The 90220 monoclonal antibody specifically binds to the beta subunit (IL-10R beta/IL-10RB/IL-10R β ) of the heteromeric IL-10 Receptor complex. This type I transmembrane glycoprotein is also known as, subunit 2 (IL-10R2) or CD210b. The IL-10 Receptor complex is formed by the binding of dimeric IL-10 to two cell surface alpha subunits of the IL-10 Receptor (CD210a/IL-10RA/IL-10R α ). This complex can then recruit two CD210b subunits resulting in the transduction of intracellular JAK/STAT pathway signals. CD210b is also a component of cytokine receptor complexes specific for IL-22, IL-26, IL-28 and IL-29. CD210b is expressed by a variety of cell types including T cells, B cells, NK cells, monocytes and macrophages.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
Specifications
| Antigen | IL-10Rbeta (CD210b) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 90220 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 421 |
| Description | IL-10R2; IL-10RB; IL-10Rβ; IL10RB; IL-10 Receptor beta; CRFB4 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human IL-10 Rβ Recombinant Protein |
| Show More |
Safety and Handling
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.