Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYAL4, Mouse, Clone: 1B10, Abnova™

Catalog No. 89106621 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-621 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-621 Supplier Abnova Corporation Supplier No. H00023553M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HYAL4.

This gene encodes a protein which is similar in structure to hyaluronidases. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. However, this protein has not yet been shown to have hyaluronidase activity. [provided by RefSeq

Sequence: FVSSDLGSYIANVTRAAEVCSLHLCRNNGRCIRKMWNAPSYLHLNPASYHIEASEDGEFTVKGKASDTDLAVMADTFSCHCYQGYEGADCREIKTADGCS

Specifications

Antigen HYAL4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HYAL4.
Formulation ascites with no preservative
Gene HYAL4
Gene Accession No. NM_012269
Gene Symbols HYAL4
Host Species Mouse
Immunogen HYAL4 (NP_036401.1,357 a.a. ∽ 456 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23553
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.