Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Kallikrein-1/KLK1, Clone: 11C7.1,

Catalog No. MABC552MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC552MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC552MI Supplier MilliporeSigma™ Supplier No. MABC552
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Kallikrein-1/KLK1 clone: 11C7.1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Kallikrein-1 (EC:3.4.21.35; UniProt P06870; also known as Glandular kallikrein 1, hK1, Kallikrein serine protease 1, Kidney/pancreas/salivary gland kallikrein, Renal/pancreas/salivary kallikrein 1, Tissue kallikrein) is encoded by the KLK1 (also known as KLKR) gene (Gene ID 3816) in human. Kallikrein-1 is a member of the kallikrein subfamily of serine proteases, members of which are involved in diverse physiological functions and many are used clinically as biomarkers for cancer. Kallikrein-1 is initially produced as a preproprotein with a signal (aa 1-18) and a propeptide (aa 19-24), cleavage of which releases the mature protein (aa 25-262). Kallikrein-1 is the major kallikrein expressed in the kidney, where it generates kinins (e.g. lysyl-bradykinin and bradykinin) and plays role in kidney function and blood pressure regulation through vasodilatory and natriuretic effects. Decreased kidney Kallikrein-1 level is reported in experimental settings of hypertension, diabetic nephropathy, high salt- and aminoglycoside-induced kidney injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein-1/KLK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 11C7.1
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P06870
Gene Symbols KLK1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the C-terminal half of human Kallikrein-1/KLK1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_002248
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.